Learn More
Fisherbrand™ Accessories for Fisherbrand™ Heating/Cooling Mini Shaker
Optional accessories to enhance the Fisher Scientific Heating/Cooling Mini Shaker
Brand:  Fisherbrand™
Additional Details : Weight : 0.70000kg
 View more versions of this product
Product Code. 15524090
  • £97.25 / Each
Estimated Shipment: 22-10-2024
to see stock.
Add to basket
Specifications
9 x 21mm Block for 20mm diameter tube; Well: 21.7mm dia X .45mm depth