Learn More
Fisherbrand™ Accessories for Fisherbrand™ Heating/Cooling Mini Shaker
Optional accessories to enhance the Fisher Scientific Heating/Cooling Mini Shaker
£85.00 - £105.04
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
15524090
|
Fisherbrand™
88861108 |
9 x 21mm Block for 20mm diameter tube; Well: 21.7mm dia X .45mm depth |
£103.00
Each |
Estimated Shipment: 24-06-2025
Log in to see stock. |
||||||
15584080
|
Fisherbrand™
88861104 |
20 x 1.5 mL micro centrifuge tube block; Well: 11.1mm dia X 39.1mm depth |
£85.00
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
15594080
|
Fisherbrand™
88861105 |
20 x 2mL micro centrifuge tube block; Well: 11.5mm dia X 38.1mm Depth |
£89.08
Each |
Estimated Shipment: 24-06-2025
Log in to see stock. |
||||||
15504090
|
Fisherbrand™
88861106 |
12 x 15mL Conical Bottom Centrifuge Tube Block; Well: 17.1mm dia X 44.5mm Depth |
£105.04
Each |
Estimated Shipment: 24-06-2025
Log in to see stock. |
||||||
15514090
|
Fisherbrand™
88861107 |
5 x 50mL Conical Bottom Centrifuge Tube Block; Well: 29mm dia X 47.6mm depth |
£101.00
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
Description
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Accessories for Fisherbrand™ Heating/Cooling Mini Shaker