missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Accessories for Fisherbrand™ Mini Tube Rotator

Optimize your Fisher Scientific™ Mini Tube Rotator with these optional accessories

Brand:  Fisherbrand™

Additional Details : Weight : 0.06000kg

 View more versions of this product

Product Code. 15564140

  • £10.07 / Each
In Stock
to see stock.
Add to basket

Description

Description

Optimize your Mini Tube Rotator with these optional accessories

  • Increase performance and flexibility
Specifications

Specifications

Paddle for 50 mL tubes
Paddle for 50mL tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers