Learn More
BRAND™ Accessories For Media Bottle
For use with media/ reagent bottle
Brand: BRAND™ 122765
This item is not returnable.
View return policy
Description
Use with media bottle (122710)Specifications
Media Bottle | |
0.45 μm | |
12 |
PES | |
Sterile | |
Vacuum Filter Funnel (0.45 μm) |