missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Accessories For Multiphor™ II Flatbed Electrophoresis System: Cooling Plate

Accessories For Multiphor™ II Flatbed Electrophoresis System

Brand:  Cytiva 18-1103-46

Product Code. 10466724

This item is not returnable. View return policy

Description

Description

  • Material: ceramic
  • Dimensions: 210 x 270mm

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Multiphor™ II Flatbed Electrophoresis System
Ceramic
27 cm
Cooling Plate
10.62 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers