Learn More
Thermo Scientific™ Accessories for HyPerforma™ Single-Use Bioreactors, Bioreactor Probe Assembly
The Thermo Scientific™ Bioreactor Probe Assembly (non-sterile) is used to package the pH and DO probes for sterilization and to aseptically connect them to the BPCs for Thermo Scientific™ HyPerforma™ Single-Use Bioreactors.
Brand: Thermo Scientific™ SH30720.01
Additional Details : Weight : 1.04326kg
Description
The Thermo Scientific™ Bioreactor Probe Assembly (non-sterile) is used to package the pH and DO probes for sterilization and to aseptically connect them to the BPCs for Thermo Scientific™ HyPerforma™ Single-Use Bioreactors.
Specifications
Non-sterile | |
Each | |
HyPerforma™ |
50 to 1000 L | |
Non-sterile |