missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Accessories for Labconco™ Fiber Glass Desiccator
Vacuum Tubing Kit allows connection of Vacuum Desiccator's valve to an accessory pump.
Brand: Labconco™ 5530600
This item is not returnable.
View return policy
Description
Powder-coated steel rack allows one Labconco Vacuum Desiccator to be placed on top of another.Order Info
Dimensions: 12L x 12 in. W
Specifications
Stacking Rack | |
Labconco Fiber Glass Desiccator | |
2.3 kg |
Epoxy-coated Steel | |
5.07 lb. |