Learn More
Labconco™ Accessories for Labconco™ Fiber Glass Desiccator
Vacuum Tubing Kit allows connection of Vacuum Desiccator's valve to an accessory pump.
£305.00
Specifications
For Use With (Equipment) Labconco Fiber Glass Desiccator
View More Specs
Products 1
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
11791379
View Documents
Labconco™
5530600
Stacking Rack
£305.00
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
Description
Description
Vacuum Tubing Kit includes 1/4 in. ID and 1/2 in. ID vacuum tubing, each 48 in. long, two each 1/4 in. and 1/2 in. tubing clamps; 1/4 in. to 1/2 in. barbed reducer. Kit allows connection of Labconco Vacuum Desiccator's 1/4 in. diameter brass needle valve to an accessory pump with 1/2 in. connector.
Order Info
Dimensions: 12L x 12 in. W
Specifications
Specifications
Labconco Fiber Glass Desiccator