Learn More
Thermo Scientific™ Accessories for Thermo Scientific™ Large Capacity Centrifuges
Complete your system with accessories for your Thermo Scientific™ large capacity centrifuge.
Brand:  Thermo Scientific™ 75007730
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15580204
  • £140.00 / Each
Estimated Shipment: 23-12-2024
to see stock.
Description
Description
Thermo Scientific™ large capacity centrifuges are designed for high-throughput processing with the flexibility to select higher capacity rotors or match existing workflows.
Accessories designed to complete your Thermo Scientific large capacity centrifuge system.
Specifications
Specifications
Drain Box
Thermo Scientific™ ™ Cryofuge 8 and 16, Sorvall™ BP 8 and 16, and Sorvall™ BIOS 16 centrifuges
Drain Box (600 x 400 x 50mm)