Learn More
Thermo Scientific™ Accessories for MaxQ™ HP Orbital Shakers
Get maximum flexibility, regardless of your application, by using these accessories for Thermo Scientific™ MaxQ™ High Performance orbital shakers.
Brand: Thermo Scientific™ 760440
Code : NEW
Additional Details : Weight : 0.05000kg
Description
Get maximum flexibility, regardless of your application, by using Thermo Scientific™ Accessories for MaxQ™ High Performance Orbital Shakers.
Specifications
Hepa Filter Replacement | |
Replacement HEPA filter | |
MaxQ HP 435, 436, 480, 481, 8000 |