Learn More
Thermo Scientific™ Accessories for Orion™ AQUAfast AQ3010 Turbidity Meter
Generate an accurate turbidity reading quickly with this reliable Turbidity Meter and its accessories.
Brand: Thermo Scientific™ AC3SIL
Code : A0
Additional Details : Weight : 0.10000kg
Description
Generate an accurate turbidity reading quickly with the Thermo Scientific™ Orion™ AQUAfast AQ3010 Turbidity Meter and its accessories. Perfect for the plant, field or lab, this simple yet powerful turbidity meter offers an impressive array of features. Use the Infrared (IR) LED light source to comply with ISO 7027 requirements and reliably measure turbidity with auto-ranging values from 0 to 1000 NTU. This IP67 waterproof meter offers a long battery life averaging about 3000 tests.
Features:
- Accessories for AQUAfast AQ3010 Turbidity Meter
Specifications
AQUAfast AQ3010 Turbidity Meter | |
Silicone Oil (1mL Bottle) And Cloth |
Safety and Handling
- Silicone Oil
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 4
Hazard Statement
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P202-Do not handle until all safety precautions have been read and understood.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : silicone oil