Learn More
Thermo Scientific™ Accessories for RT Basic Series and RT Touch Series Magnetic Stirrers
Use these accessories to optimize performance of your Thermo Scientific™ RT Basic Series Magnetic Stirrers.
Brand:  Thermo Scientific™ 88880146
 View more versions of this product
Product Code. 15238904
  • £13.05 / Each
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use these Thermo Scientific™ Accessories for RT Basic Series Magnetic Stirrers to optimize performance of your stirrer.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
RT Basic Series and RT Touch Series Magnetic Stirrers
Plate Cover
Non-slip silicone plate cover for items 88880011 and 88880012 (White color, Silicone 220mm dia.)
96-Well Plates, 384-Well Plates
Silicone