Learn More
Sartorius Accessories for Vivacell Models
Accessories for various Vivacell units
£80.96 - £119.00
Product Code | Brand | For Use With (Equipment) | For Use With (Application) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | For Use With (Application) | Includes | Price | Quantity & Availability | |||
10663382
|
Sartorius
VCA005 |
For Vivacell 20 and 70; For Pressure-fuge head | Charge Valve | Not Applicable |
£119.00
Each |
In Stock
Log in to see stock. |
|||
10059730
|
Sartorius
VCA009 |
Vivacell 250 | Maintenance Kit | Sample reservoir; filtrate container; “O” ring seals | N/A | ||||
Description
- Use charge value with pressure-fuge head
- Maintenance Kit includes sample reservoir, filtrate container and “O” ring seals
Specifications
Accessory for Vivacell Model |