Learn More
Merck Accessories for XX6702500 FilterJet Solvent Dispenser
Accessories for filter holders for sample preparation.
Brand:  Merck XX40025RK
245.68 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : 72
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10218300
  • £281.00 / Each
Estimated Shipment: 20-09-2024
to see stock.
Add to basket
Description
Description
Accessories for filter holders for sample preparation.
Specifications
Specifications
Filterjet Solvent Dispenser
PTFE Locking Nut Washers, PTFE O-Rings, Stainless Steel Support Screen
Replacement Parts Kit