missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories for XX6702500 FilterJet Solvent Dispenser

Accessories for filter holders for sample preparation.

Brand:  Merck XX6702507

73.37 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 72

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10758821

  • £78.75 / Each
Estimated Shipment: 20-09-2024
to see stock.
Add to basket

Description

Description

Accessories for filter holders for sample preparation.

  • Made from Series 300 stainless-steel
  • Adapter (XX6702507) is a replacement part for Filterjet™ Solvent Dispenser and is included in Connector Kit for ZHE Hazardous Waste Filtration System

Specifications

Specifications

NPTM to tube
Stainless Steel
183 cm
0.25 in.
0.63 cm
Filterjet Solvent Dispenser
Tube Adapter, 1/4 in. NPTM to 1/4 in. Tube
0.25 in.
6.3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers