missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories for YT31042HW Hazardous Waste Filtration System, 142 mm

Accessories for filter holders for sample preparation.

Brand:  Merck YT3014257

1355.08 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10334951

  • £1555.00 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Accessories for filter holders for sample preparation.

YT3014257 Underdrain Support, PTFE-coatedYT310RAHW Rotary Agitator
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
142 mm hazardous waste filtration system
Non-sterile
Underdrain Support
PTFE Coated Stainless Steel
Environmental Test Kits or Supplies
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers