missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Accessories for YT31042HW Hazardous Waste Filtration System, 142 mm
Accessories for filter holders for sample preparation.
Brand: Merck YT310RAHW
6158.47 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Includes: Accessory clamps for 1- and 2.5-liter bottle extractions
Description
Accessories for filter holders for sample preparation.
YT3014257 Underdrain Support, PTFE-coatedYT310RAHW Rotary AgitatorOrder Info
Dimensions: 21L x 26 1/4W x 24 3/8 in.H (53 x 66.6 x 61.9cm)
Specifications
Not Autoclavable | |
Non-sterile | |
Environmental Test Kits or Supplies |
Rotary Agitator | |
Accessory clamps for 1 L and 2.5 L bottle extractions |