Learn More
IKA™ R 1303 Dissolver Stirrer
Draws material to be mixed from the top and the bottom with radial flow. IKA R 1303 Dissolver Stirrer creates high turbulence and high shearing forces with particle reduction. For use at medium to high speeds.
Brand:  IKA™ 0002746700
Product Code. 10005220
  • £496.00 / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Radial flow, for drawing the material to be mixed from the top and the bottom. High turbulence, high shearing forces. Particle reduction. Used at medium to high speeds. Stirrer Ø: 42 mm Shaft Ø: 8 mm Shaft length: 350 mm
Specifications
Specifications
1.7 in.
42 mm
13.8 in.
2000 rpm
0.3 in.
8 mm
350 mm
Stirring Shaft