Learn More
IKA™ Accessory Stirring Shafts for SteadyStir™ and Overhead Stirrers
Generate axial flow in the vessel with these stirring shafts. IKA™ Accessory Stirring Shafts for SteadyStir™ and IKA™ Overhead Stirrers are designed for medium to high speeds.
£172.00 - £650.00
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
10568881
|
IKA™
0000757700 |
R 1375 |
£196.00
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
10767381
|
IKA™
0000513500 |
R 1300 |
£290.00
Each |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
10201961
|
IKA™
0002387900 |
R 1302 |
£650.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10160161
|
IKA™
0002332900 |
R 1311 |
£220.00
Each |
Estimated Shipment: 09-09-2024
Log in to see stock. |
|||||
10642781
|
IKA™
0000741300 |
R 1345 |
£218.78
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10058130
|
IKA™
0000756900 |
R 1352 |
£183.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
10435261
|
IKA™
0000741000 |
R 1342 |
£172.00
Each |
Estimated Shipment: 22-07-2024
Log in to see stock. |
|||||
Description
Four-Bladed Propeller Stirrers
- Standard stirring element
- For drawing material to be mixed from top to bottom
- Local shearing force
- Generates axial flow
- Use at medium to high speeds
- Flow efficient design
- For drawing material to be mixed from top to bottom of vessel
- Minimum shearing
- Medium to high speeds
- For drawing material to be mixed from above
- Axial flow
- Minimal danger of injury
- Medium to high speeds
- Radial flow for drawing material to be stirred from top and bottom of vessel
- High turbulance, high shearing forces
- For particle reduction
- Minimum and high speeds
- Blades open with increasing speed
- For stirring in round vessels with narrow necks
- Medium to high speeds
- Tangential flow, minimal turbulance, good heat exchange, gentle treatment of product
- Low to medium speeds
- Ideal stirrers for medium-to-high-viscosity fluids
- Tangential flow, high shearing rate at edges, minimal deposits
- Low speeds
Key to IKA models in ordering table below: RW16 Basic Stirrer (RW16), Eurostar Power Basic (Basic), Eurostar Power Control-Visc (Control-Visc), Eurostar Power Control-Visc P4 (P4), Eurostar Power Control-Visc P7 (P7), Eurostar Power Control-Visc 6000 (CV6000)
† PTFE-coated