missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ PepMap™ 100 C18 HPLC Columns

Product Code. 17294434 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
5 mm
15 mm
20 mm
50 mm
150 mm
250 mm
500 mm
750 mm
Diameter (Metric):
0.05 mm
0.075 mm
0.1 mm
0.2 mm
0.3 mm
0.75 mm
75 μm
1 mm
Particle Size:
2 μm
3 μm
5 μm
Type:
Analytical Column
Analytical Column, NanoViper
Analytical nano Column
Column
Trap Cartridge
Trap Column
Unit Size:
Each
Pack of 2
Pack of 5

Product Code. 17294434

Brand: Thermo Scientific™ 164750

Only null left
Add to basket
Add to basket

Use for high-resolution tryptic, natural, and synthetic peptide analyses. Its high loading capacity is exceptional for low abundant peptides in complex proteomic samples.

Feature high loading capacity for exceptional suitability in the analysis of low abundant peptides in complex proteomics samples with Thermo Scientific™ PepMap™ 100 C18 LC Columns. PepMap LC columns offer lengths targeting fast or high resolution peptide separation.

  • High resolution in protein identification, biomarker discovery, and systems biology
  • Particular loadability allows highest sensitivity in LC/MS
  • Designed for TFA-free LC-MS, minimizing ion-suppression effects
  • Optimized for coupling to ESI/MS and MALDI-MS
  • Highest column-to-column reproducibility
  • Easy-to-use, cutting-edge miniaturized HPLC
  • Preassembled with nanoViper column inlet fittings for easy column installation

Key advantages of nanoViper fittings

  • Ease of use, no sleeve for small capillaries
  • Tool-free assembly
  • No column damage from overtightening: capable of withstanding pressures up to 1000 bars and is compatible with third-party valves and unions
  • No experimental failure due to bad connections

Ideal for high-resolution peptide mapping of tryptic, natural, and synthetic peptides as well as LC-MS/MS peptide mapping for protein identification, biomarker discovery, and systems biology.

  • High loading capacity
  • Use for the analysis of low abundant peptides often found in complex proteomics samples
  • Select the column length and I.D. for your throughput, sensitivity, and amount of sample available requirements
  • Choose from 50, 150, 250, 500mm lengths and 50, 75, 300 and 1000μm I.D.
  • Superior protein and peptide separations
  • Trifluoroacetic acid (TFA)-free LC/MS minimizes suppression effects for enhanced MS sensitivity
  • Outstanding separation efficiency
  • Superior resolution and recovery
  • Easy installation with nanoViper™ Fingertight Fittings

PepMap nanocolumns are also available pre-assembled with Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fitting for a dead-volume-free connection to 1000 bar.

  • Requires no tools
  • Provides virtually zero-dead-volume connections
  • Withstands pressures up to a 1000 bar

About Nano, capillary, and micro Acclaim PepMap columns

Nano (75μm), capillary (300μm) and micro (1000μm) columns are ideal for protein and peptide analyses. Sample amount is often limited and smaller I.D. columns can increase in sensitivity in sample-limited situations. While nano LC columns are often used in proteomics, the concentrating effects of capillary and microcolumns are beneficial for metabolomics or for facilitating LC-MS interfacing. Thermo Scientific Acclaim PepMap columns are offered in nano, capillary, and microcolumn in a variety of chemistries for peptides and protein analysis.

Use PepMap Trap columns for desalting and preconcentration of peptides before LC separation with MS detection. The trap columns provide the highest efficiencies for one-dimensional peptide mapping experiments and 2D-LC analyses.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Trap Column, Nanoviper
Diameter (Metric) 0.1 mm
For Use With UltiMate 3000 RSLC Nano System
Length (Metric) 150 mm
Max. Pressure 7250 psi (500 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 100 Å
Quantity Pack of 2
Surface Area 450 m2/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line nanoLC
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 15%
Column Type Peptide Analysis
Endcapped Yes
Phase Reversed Phase
Type Trap Column
Bed Length 20 mm
Mehr anzeigen Weniger anzeigen

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt