missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ C30 HPLC Columns

Use for superior separation of hydrophobic structural isomers.

Brand:  Thermo Scientific™ 075725

Code : 16

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11362023

  • £738.12 / Each
Estimated Shipment: 01-10-2024
to see stock.
Add to basket

Description

Description

For separations of hydrophobic, structurally related isomers, Thermo Scientific™ Acclaim™ C30 LC reversed-phase columns provide superior results. The covalent modification of C30 alkyl silane on ultrapure, spherical, porous silica gel gives these columns high shape selectivity. They are fully compatible with various aqueous buffers, useful for a broad application range of analytes, and offer more flexibility in method development.

High Shape Selectivity for the Separation of Structurally Related Isomers
  • Unique selectivity complementary to other reversed-phase columns
  • High efficiency and rugged packing
  • Extremely low bleed, fully compatible with MS
  • Stable between pH 2 and 8
  • Available in 3 and 5μm particle sizes and 4.6, 3.0, and 2.1mm diameters, with an average pore diameter of 200Å
About the Acclaim Family of HPLC Columns

Achieve the high standards set by modern HPLC and LC/MS methods with the Acclaim family of reversed-phase columns. These silica-based columns with advanced column bonding and endcapping technologies provide complementary selectivity, high column efficiencies, and ultra-high resolution for a wide variety of applications.

  • Broad spectrum of stationary phases, including conventional C8 and C18 phases, polar embedded phases, and specialty phases including wide-pore, are available to meet your specific application needs
  • Performance tested to ensure high quality and reliability for pharmaceutical, biopharmaceutical, environmental, food, and other industrial chromatographic separations
Specifications

Specifications

Analytical Column
LC/MS
8500 psi (586 bar)
2 to 8
200 m²/g
L62
Acclaim
13%
Proprietary
2.1 mm
150 mm
3 μm
200 Å
60°C
C30
Spherical, Fully Porous, Ultrapure Silica
Reversed Phase
Reversed Phase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.