missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acclaim™ Trinity Q1 LC Columns

Use to separate the herbicides diquat and paraquat.

Brand:  Thermo Scientific™ 079717

704.46 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 15501561

  • £817.77 / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Simultaneously separate the herbicides diquat and paraquat with Thermo ScientificTM AcclaimTM Trinity Q1 LC Columns. These tri-mode (WCX, WAX, RP) columns offer unmatched high-resolution and high-throughput trace analysis through LC-MS/MS and LC-UV methods. The stationary phase, based on Nanopolymer Silica Hybrid (NSH) technology, provides multiple retention mechanisms, including reversed-phase, anion-exchange, and cation-exchange, for maximum flexibility in method development. Easily optimize the selectivity to accelerate chromatographic separations and increase productivity.

Superior Performance for Diquat and Paraquat Analysis
  • Distinctive spatial separation of anion-exchange and cation-exchange regions for unparalleled chromatographic performance
  • Adjustable selectivity using mobile phase buffer concentration, pH, and solvent content
  • Quick separation of hydrophilic ionic and ionizable analytes without ion-pairing reagent
  • Ideal selectivity for simultaneous separation of basic, neutral, and acidic analytes
  • Compatibility with mass spectrometry detection, which significantly improves sensitivity and provides conformation at the same time
  • Available in 3.0μm particle size and 2.1 and 3.0mm diameters, in 50mm length

About the Family of Acclaim Specialty HPLC Columns

Acclaim Specialty Columns have been developed, manufactured and tested to meet the separation needs of specific applications. These columns are based on novel and unique chemistries and provide superior resolution with ease-of-use.

  • Use-tested to guarantee consistent separations
  • Ideal selectivity for separating specific target analytes
  • Excellent peak shapes
  • Performance tested to ensure high quality and reliability for specific chromatographic separations
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Each
3 μm
300Å
6000 psi
2.1 mm
Analytical Column
Yes
2.5 to 7.5
100 mm
100 m2/g
2.1 mm
40°C
Specialty / Application Specific
Acclaim
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number001508 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number079717
Lot Number001509 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number079717
Lot Number001546 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number079717
Lot Number001561 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number079717
Lot Number001563 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number079717
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Acclaim™ Trinity Q1 LC Columns >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.