missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ accumet™ AE6 Portable pH/ORP Meter

The AE6 Portable pH Meters are ideal for quick, accurate conductivity or TDS checks in laboratories, field and schools.

Brand:  Fisherbrand™ 13-636-AE6

475.00 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.


 View more versions of this product

Product Code. 15523653

  • £500.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: pH/Temp electrode
This item is not returnable. View return policy

Description

Description

Fisherbrand accumet AE6 Portable pH Meter, Includes: pH/Temp electrode, Accuracy: +/- 0.01pH, +/-0.5 deg.C, +/- 1mV, Range: -1000 to 1000 mV, 0 to 14, Resolution: 0.01, 1mV, 0.1 deg.C, Battery Type: 4 x AAA

  • Multipoint calibration (up to 5 points) with auto-buffer recognition
  • Choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
  • For analyzing water in pools and spas, lithographic processes, boiler and cooling tower water analysis, environmental and agricultural purposes, and all types of quality assurance and water quality testing
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Hold function momentarily freezes reading for easy viewing and recording
  • Auto-power off saves battery power after 17 minutes of non-use
  • Self-diagnostic with message codes for easy troubleshooting
  • Large custom LCD provides optimum viewing, even at a distance
  • Rubber boot shields protect from accidental knocks and feature sturdy, built-in stand for easy benchtop operation
  • Optional rugged carrying case provides handy on-site meter calibration and measurement
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AE6 pH/ORP Meter, Rubber Armor/Stand, 3-in-1 pH/Temperature Combination Electrode ECFC7352901B and pH Carrying Kit
±1
Water Quality Testing
0 to 14
-1000 to +1000 mV
1 to 5
0.1°C
AAA
±0.5°C
±0.01
1.68, 4.01, 7, 10.01, 12.45
0.01
1 mV
0°C to 100°C
Automatic/Manual (0°C to 100°C)
Portable Meter
Product Suggestions

Product Suggestions

Videos

Videos

  • Fisherbrand accumet AE6 pH and AE6C conductivity meters

SDS
Documents

Documents

Brochures

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ accumet™ AE6 Portable pH/ORP Meter > AE6 Portable pH Meter, Includes: AE6 pH/ORP Meter, Rubber Armor/Stand, 3-in-1 pH/Temperature Combination Electrode ECFC7352901B and pH Carrying Kit; Battery Type: AAA; Accuracy (mV): ±1; Range (mV): -1000 to +1000 mV

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.