missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Basic AB315 Benchtop Laboratory pH/mV Meter
Ideal for daily pH testing of your routine laboratory samples, the accumet™ Basic AB315 Benchtop Meter provides essential functions needed for common pH measurements without complicated functions, all at a budget-friendly price.
£490.00 - £789.48
Specifications
Accuracy (mV) | ±0.2 or ±0.05%, Whichever is Greater |
---|---|
Accuracy (pH) | ±0.01 |
Accuracy (Temperature) | ±0.3°C (±0.5°F) |
Data Management Details | Printed or CSV Format |
Outputs | Port for Computer or Printer |
Product Code | Brand | Description | Includes | Certifications/Compliance | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Includes | Certifications/Compliance | Price | Quantity & Availability | |||
17388949
|
Fisherbrand™
|
pH/mV Meter with Electrode Stand | Electrode Stand, Power Adapter | CE, TUV 3-1, FCC Class A, Without NIST-traceable Calibration Certificate |
£490.00
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
17398949
|
Fisherbrand™
|
pH/mV Meter with pH/ATC Epoxy-body Double Junction Gel-filled Electrode | pH/ATC Electrode, Electrode Stand, Power Adapter | CE, TUV 3-1, FCC Class A, Without NIST-traceable Calibration Certificate |
£710.00
Each |
In Stock
Log in to see stock. |
|||
17308959
|
Fisherbrand™
|
pH/mV Meter with pH Glass-body Double Junction Refillable Electrode | pH Electrode, ATC Probe, Electrode Stand, Power Adapter | CE, TUV 3-1, FCC Class A, Without NIST-traceable Calibration Certificate |
£789.48
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||
Description
- Large, backlit display simultaneous shows pH, mV and temperature readings
- One to five point pH calibration with automatic buffer recognition of four pH buffer sets
- One point relative mV calibration (RmV) for ORP measurements
- One point ATC temperature probe calibration
- Calibration due alarm, selectable 1 to 60 hr., or off
- Automatic or manual temperature compensation
- Ready icon indicates measurement stability
- Electrode condition icon indicates electrode status 500 data set storage with time/date stamp
- Non-volatile meter memory preserves settings and data in event of power loss
- Meter-attached stand facilitates easy electrode movement into and out of samples
Specifications
±0.2 or ±0.05%, Whichever is Greater | |
±0.3°C (±0.5°F) | |
Port for Computer or Printer | |
pH, Relative mV, Temperature | |
0.01, 0.1 | |
±2000 mV | |
1 to 5 (pH), 1 (Relative mV), 1 (Temperature) | |
23°F to 221°F | |
5 in. LCD | |
Non-volatile | |
Automatic, Manual | |
IP54 | |
3 Years | |
±2000 mV | |
Automatic, Manual | |
100/240 V |
±0.01 | |
Printed or CSV Format | |
pH with mV and Temperature, Relative mV with mV and Temperature | |
-2 to 18 | |
USA: 2, 4.01, 7, 10.01, 12; NIST: 1.68, 4.01, 6.87, 9.18, 12.46; DIN: 1.09, 3.06, 4.65, 6.79, 9.23, 12.75; FSCI: 1, 3, 6, 8, 10, 13 | |
1 mV, 0.1 RmV | |
-5°C to +105°C | |
0.1°C (0.1°F) | |
Yes | |
500 Points | |
BNC, ATC | |
Universal Power Adapter | |
AB315 | |
Indicator Icons | |
1 Channel | |
50/60 Hz |