missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ FE150 Benchtop pH Meter
Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price.
Brand: Fisherbrand™ S35924
309.42 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet FE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.
- One to five point pH calibration
- Automatic buffer recognition with two pH buffer sets
- pH slope and offset display
- One point mV calibration
- One point ATC temperature probe calibration
- Automatic or manual temperature compensation
- Ready icon to indicate measurement stability
- Electrode condition icon to indicate status
- 100 data set storage with non-volatile memory
Specifications
Meter, Double Junction pH Electrode, Temperature Probe and Stand | |
Recorder (±2000 mV) | |
0.01 | |
0.1 mV (±199.9 mV); 1 mV Beyond ±200 mV | |
0°C to 100°C | |
Offset in 0.1°C Increments; Offset Range: ±5°C | |
BNC, 2.5 mm Phono Plug, Reference Pin Tip, Ground | |
Automatic or Manual (0°C to 100°C) | |
AC/DC 9 V Adapter (110/220 VAC, 50/60 Hz) |
±0.2 ±199.9 ±2 Beyond ±200 | |
0 to 14 | |
±1999 mV | |
USA: pH 1.68, 4.01, 7.00, 10.01, 12.45; NIST: pH 1.68, 4.01, 6.86, 9.18, 12.45 | |
0.1°C | |
100 Data Sets | |
3 Year Meter Warranty | |
Bio Set |