Learn More
Fisherbrand™ accumet™ FE150 Benchtop pH Meter
Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price.
Brand: Fisherbrand™ S35925
354.17 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet FE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.
- One to five point pH calibration
- Automatic buffer recognition with two pH buffer sets
- pH slope and offset display
- One point mV calibration
- One point ATC temperature probe calibration
- Automatic or manual temperature compensation
- Ready icon to indicate measurement stability
- Electrode condition icon to indicate status
- 100 data set storage with non-volatile memory

Specifications
Meter, 13-620-FE6 3-in-1 Single-Junction Gel pH/ATC Electrode, and Stand | |
±0.2 ±199.9 ±2 Beyond ±200 | |
Recorder (±2000 mV) | |
0.01 | |
0.1 mV (±199.9 mV); 1 mV Beyond ±200 mV | |
0°C to 100°C | |
Offset in 0.1°C Increments; Offset Range: ±5°C | |
BNC, 2.5 mm Phono Plug, Reference Pin Tip, Ground | |
Automatic or Manual (0°C to 100°C) | |
AC/DC 9 V Adapter (110/220 VAC, 50/60 Hz) |
±0.3°C | |
±0.01 | |
0 to 14 | |
±1999 mV | |
1 to 5 with Automatic Buffer Recognition (ABR) | |
0.1°C | |
100 Data Sets | |
3 Year Meter Warranty; 6 Month Electrode Warranty | |
3-in-1 Set |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
34Hours
0Minutes
17Seconds
30 -
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.