missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ FE150 Benchtop pH Meter
Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price.
£302.00 - £433.00
Specifications
Accuracy (mV) | ±0.2 ±199.9 ±2 Beyond ±200 |
---|---|
Outputs | Recorder (±2000 mV) |
Range (pH) | 0 to 14 |
Range (mV) | ±1999 mV |
Resolution (mV) | 0.1 mV (±199.9 mV); 1 mV Beyond ±200 mV |
Product Code | Brand | Includes | Type | Calibration Points | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Type | Calibration Points | Price | Quantity & Availability | |||
17721739
|
Fisherbrand™
|
Meter, Electrode Stand, Universal Power Adapter (90 to 240 V, 50/60 Hz) | Standard Set | 1 to 5 with Automatic Buffer Recognition (ABR) |
£302.00
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||
17731739
|
Fisherbrand™
|
Meter, Temperature Probe and Stand | Education Set | USA: pH 1.68, 4.01, 7.00, 10.01, 12.45; NIST: pH 1.68, 4.01, 6.86, 9.18, 12.45 |
£387.00
Each |
In Stock
Log in to see stock. |
|||
17741739
|
Fisherbrand™
|
Meter, Double Junction pH Electrode, Temperature Probe and Stand | Bio Set | USA: pH 1.68, 4.01, 7.00, 10.01, 12.45; NIST: pH 1.68, 4.01, 6.86, 9.18, 12.45 |
£433.00
Each |
In Stock
Log in to see stock. |
|||
17751739
|
Fisherbrand™
|
Meter, 13-620-FE6 3-in-1 Single-Junction Gel pH/ATC Electrode, and Stand | 3-in-1 Set | 1 to 5 with Automatic Buffer Recognition (ABR) |
£409.70
Each |
In Stock
Log in to see stock. |
|||
Description
Designed for routine pH measurements in the lab with convenience in mind – simple functionality, stored data sets in a non-volatile memory and meter-attachable electrode stand. The accumet FE150 pH meters offer efficient sample reading and recording with flexible display options to show display both pH and temperature or mV and temperature simultaneously.
- One to five point pH calibration
- Automatic buffer recognition with two pH buffer sets
- pH slope and offset display
- One point mV calibration
- One point ATC temperature probe calibration
- Automatic or manual temperature compensation
- Ready icon to indicate measurement stability
- Electrode condition icon to indicate status
- 100 data set storage with non-volatile memory
Specifications
±0.2 ±199.9 ±2 Beyond ±200 | |
0 to 14 | |
0.1 mV (±199.9 mV); 1 mV Beyond ±200 mV | |
0.1°C | |
100 Data Sets | |
AC/DC 9 V Adapter (110/220 VAC, 50/60 Hz) |
Recorder (±2000 mV) | |
±1999 mV | |
0°C to 100°C | |
Offset in 0.1°C Increments; Offset Range: ±5°C | |
BNC, 2.5 mm Phono Plug, Reference Pin Tip, Ground |