Learn More
Invitrogen™ AccuPrime™ SuperMix I
AccuPrime SuperMix I is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification.
£642.00 - £2185.00
Specifications
Gel Compatibility Low
Fidelity (vs. Taq) 2X
View More Specs
Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
10297522
View Documents
Invitrogen™
12342028
1000 Reactions This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10564233
View Documents Promotion Details
Invitrogen™
12342010
200 Reactions
£642.00
Each
Description
Description
AccuPrime SuperMix I is a ready-to-use mixture of DNA polymerase, accessory proteins, salts, magnesium, and dNTPs for efficient PCR amplification. All you have to do is add template and primers, reducing set-up time by half. For accurate PCR performance, AccuPrime SuperMix I sets the standard. You'll amplify your DNA targets cycle after cycle, eliminating troubleshooting and rework.
In addition to a powerful DNA polymerase, AccuPrime SuperMix I includes Platinumanti-DNA polymerase antibodies that inhibit polymerase activity, providing an automatic hot-start, and a thermal stable accessory protein that enhances specific primer-template hybridization during every cycle of PCR. This unique combination allows for success, even under previously suboptimal conditions.
AccuPrime SuperMix I is designed for amplification of genomic DNA amplicons (> 200 bp), plasmid DNA, or cDNA templates. Alternatively, AccuPrime SuperMix II is designed to amplify a broader range of amplicon sizes up to 4 kb.
• Built-in hot start
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
Low
2X