Learn More
Fisherbrand™ accuQuatro U Pack Cartridge
Choose the cartridge for your feed water type. Fisher Scientific™ AccuQuatro Cartridges are available for reverse osmosis, deionized, or low TOC feed water types.
Brand:  Fisherbrand™
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 15409719
  • £855.00 / Each
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
Description
Description
  • Minimal dead water volume
  • Production of ion-free and organic-free ultrapure water
  • Choices of ultrapurification cartridges for feed water types
Specifications
Specifications
Accu100
AccuQuatro U pack Cartridge Kit with 0.2μm final filter, DI Feed