missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACE Mouse ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EMACE

Code : NEW

Additional Details : Weight : 0.00400kg

 View more versions of this product

Product Code. 15415183

  • £504.00 / 1 set
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Description

Description

The Mouse Angiotensin Converting Enzyme (Ms ACE) ELISA quantitates Ms ACE in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms ACE. Principle of the method The Mouse ACE solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287 bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes - the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined.
Specifications

Specifications

P09470
0.12 ng/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
11421
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Cardiovascular Research
2°C to 8°C
Mouse
4 hr. 45 min.
0.123-30 ng/mL
0.12ng/ml to 30ng/ml
Biotin
Proteases, Peptidases & Inhibitors
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN-gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF.
Mouse
Colorimetric Microplate Reader
AW208573,CD143
<10%
HRP
RUO
Plasma, 1 μL; Serum, 1 μL; Supernatant, 100 μL
RP23-100A9.1, AW208573, CD143
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers