Learn More
ACE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21115-1-AP-150UL
 View more versions of this product
Product Code. 16820725
  • £349.00 / 150µL
Explore more special offers
Description
Description
This antibody is specific to ACE2.
Angiotensin-converting enzyme 2 (ACE2) plays a central role in vascular, renal, and myocardial physiology. In contrast to its homolog ACE, ACE2 expression is restricted to heart, kidney, and testis. Recently. ACE2 has also been shown to be a functional receptor of the SARS coronavirus. The normal function of ACE2 is to convert the inactive vasoconstrictor angiotensin I (AngI) to Ang1-9 and the active form AngII to Ang1-7, unlike ACE, which converts AngI to AngII. While the role of these vasoactive peptides is not well understood, lack of ACE2 expression in ace2-/ace2- mice leads to severely reduced cardiac contractility, indicating its importance in regulating heart function.
Specifications
Specifications
ACE2
Polyclonal
Unconjugated
ACE2
ACE related carboxypeptidase, ACE2, ACEH, Angiotensin converting enzyme 2, Metalloprotease MPROT15
Rabbit
Antigen Affinity Chromatography
RUO
302668, 59272, 70008
-20°C
Liquid
Immunoprecipitation, Western Blot
0.43 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5EGZ1, Q8R0I0, Q9BYF1
ACE2
ACE2 Fusion Protein Ag15455
150 μL
Primary
Human, Mouse, Rat