missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals ACES, 99+%, for biochemistry

ACES, >-99%, C4H10N2O4S, CAS Number-7365-82-4, N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine, 1kg, 0.02 max. at 280nm, 0.04 max. at 260nm, (5% aq. soln.), (5% aq. soln.) at 0nm, 99+%, 100.0, 99.0, CHEBI:39062, 100ppm max., White, 230-908-4, 182.19, Biochemical

£64.80 - £385.00

Chemical Identifiers

CAS 7365-82-4
Molecular Formula C4H10N2O4S
Molecular Weight (g/mol) 182.194
MDL Number MFCD00008030
InChI Key DBXNUXBLKRLWFA-UHFFFAOYSA-N
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine
PubChem CID 81832
ChEBI CHEBI:39062
IUPAC Name 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NCC(=O)N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10183531
View Documents
Thermo Scientific Chemicals
327671000
100 g Plastic bottle
£64.80
100g
Please to purchase this item. Need a web account? Register with us today!
10741745
View Documents
Thermo Scientific Chemicals
327672500
250 g Plastic bottle
£130.00
250g
Please to purchase this item. Need a web account? Register with us today!
10117813
View Documents
Thermo Scientific Chemicals
327670010
1 kg Plastic bottle
£385.00
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for biochemistry
Specifications

Chemical Identifiers

7365-82-4
182.194
DBXNUXBLKRLWFA-UHFFFAOYSA-N
81832
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
C4H10N2O4S
MFCD00008030
N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine
CHEBI:39062
C(CS(=O)(=O)O)NCC(=O)N

Specifications

(5% aq. soln.), (5% aq. soln.) at 0nm, 0.02 max. at 280nm, 0.04 max. at 260nm
7365-82-4
99.0
White
Crystals
Authentic
H2NCOCH2NHCH2CH2SO3H
15, 37
Solubility in water: soluble (approx. 40g/L at 20 °C).
C(CS(=O)(=O)O)NCC(=O)N
182.194
CHEBI:39062
≥99%
ACES
(5% aq. soln.)
270.0°C to 295.0°C
100.0
3.6 to 4.4 (1% soln. at 25°C)
99+%
C4H10N2O4S
MFCD00008030
N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine
DBXNUXBLKRLWFA-UHFFFAOYSA-N
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
81832
182.19
Biochemical
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 230-908-4

missing translation for 'tsca' : TSCA

Product Identifier
  • ACES

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only