missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals ACES, 99%, extra pure

£44.70 - £274.00

Chemical Identifiers

CAS 7365-82-4
Molecular Formula C4H10N2O4S
Molecular Weight (g/mol) 182.194
MDL Number MFCD00008030
InChI Key DBXNUXBLKRLWFA-UHFFFAOYSA-N
Synonym N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine
PubChem CID 81832
ChEBI CHEBI:39062
IUPAC Name 2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
SMILES C(CS(=O)(=O)O)NCC(=O)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10400121
View Documents Promotion Details
Thermo Scientific Chemicals
228000250
25 g
£44.70
25g
Please to purchase this item. Need a web account? Register with us today!
10331564
View Documents Promotion Details
Thermo Scientific Chemicals
228001000
100 g
£116.00
100g
Please to purchase this item. Need a web account? Register with us today!
10782495
View Documents Promotion Details
Thermo Scientific Chemicals
228002500
250 g
£274.00
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Chemical Identifiers

7365-82-4
182.194
DBXNUXBLKRLWFA-UHFFFAOYSA-N
81832
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
C4H10N2O4S
MFCD00008030
N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine
CHEBI:39062
C(CS(=O)(=O)O)NCC(=O)N

Specifications

(c=5, H2O), (c=5, H2O) at 0nm, 0.02 max. at 280nm, 0.04 max. at 260nm
7365-82-4
100.0
3.6 to 4.4 (1% soln.)
99%
C4H10N2O4S
MFCD00008030
15, 37
DBXNUXBLKRLWFA-UHFFFAOYSA-N
2-[(2-amino-2-oxoethyl)amino]ethanesulfonic acid
81832
182.19
Extra Pure
(c=5, H2O)
98.5
White
Crystalline Powder
Authentic
H2NCOCH2NHCH2CH2SO3H
Glass bottle
N-(2-Acetamido)-2-aminoethanesulfonic acid, N-(Carbamoylmethyl)taurine
C(CS(=O)(=O)O)NCC(=O)N
182.194
CHEBI:39062
≥98.5%
ACES, Extra Pure
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ACES

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only