missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Acetal Valved Quick-Disconnect Hose Barb Insert

Brand:  Cole-Parmer™ 06360-43

 View more versions of this product

Product Code. 11735149

  • £15.60 / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Hose Barb Insert (Valved)
Acetal
Fits 0.18 in. Tubing I.D.
Autoclavable
4.8 cm
1 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers