missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 11484980
Click to view available options
Quantity:
100 mL
500 mL
4 x 500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Acetaldehyde is used in the production of drugs, perfumes, explosives, lacquers and varnishes, photographic chemicals, phenolic and urea resins, rubber accelerators and antioxidants. It is used as an intermediate for the synthesis of acetic acid, ethyl acetate, acetic anhydride, pentaerythritol, peracetic acid, chloral and glyoxal. As a solvent, it is used in rubber production, tanning of leather and in the paper industry. It is used as a fruit and fish preservative, as a flavoring agent, as a denaturant for alcohol and in composition of fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-07-0
Molecular Formula C2H4O
Molecular Weight (g/mol) 44.053
MDL Number MFCD00006991
InChI Key IKHGUXGNUITLKF-UHFFFAOYSA-N
Synonym ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
PubChem CID 177
ChEBI CHEBI:15343
IUPAC Name acetaldehyde
SMILES CC=O

Specifications

Melting Point -128°C
Color Colorless
Density 0.780 g/mL
Boiling Point 20.8°C
Flash Point -27°C (-17°F)
Odor Pungent
Refractive Index 1.3316
Quantity 500 mL
UN Number UN1089
Beilstein 505984
Sensitivity Air Sensitive
Merck Index 14,39
Solubility Information Miscible with water,ethanol,ether,benzene,toluene,xylene,turpentine,acetone and gasoline.Slightly miscible with chloroform.
Formula Weight 44.06
Percent Purity 98.5%
Physical Form Liquid
Chemical Name or Material Acetaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'