Learn More
Acetaldehyde ethyl propargyl acetal, 98%, Thermo Scientific™
£19.10 - £55.20
Chemical Identifiers
CAS | 18669-04-0 |
---|---|
Molecular Formula | C7H12O2 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD00066344 |
InChI Key | QKBBTQJLUGADEG-UHFFFAOYSA-N |
PubChem CID | 86774 |
IUPAC Name | 3-(1-ethoxyethoxy)prop-1-yne |
SMILES | CCOC(C)OCC#C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10025493
|
Thermo Scientific Acros
390640010 |
1 g | Glass bottle |
£19.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10732084
|
Thermo Scientific Acros
390640050 |
5 g | Glass bottle |
£55.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
18669-04-0 | |
128.17 | |
QKBBTQJLUGADEG-UHFFFAOYSA-N | |
3-(1-ethoxyethoxy)prop-1-yne |
C7H12O2 | |
MFCD00066344 | |
86774 | |
CCOC(C)OCC#C |
Specifications
18669-04-0 | |
100.0 | |
0.8930g/mL | |
Authentic | |
C7H12O2 | |
MFCD00066344 | |
3-1-ethoxyethoxy prop-1-yne, acetaldehyde ethyl propargyl acetal, 1-propyne, 3-1-ethoxyethoxy, 1-ethoxy-1-prop-2-yn-1-yloxy ethane, 3-1-ethoxyethoxy propyne, 1-ethoxyethoxy-2-propyne, 1-1-ethoxyethoxy-2propyne, 1-1-ethoxyethoxy-2-propyne, 5-methyl-4,6-dioxa-1-octyne, 1-propyne,3-1-ethoxyethoxy | |
CCOC(C)OCC#C | |
128.17 | |
128.17 | |
Liquid |
97.5 | |
Colorless to Yellow | |
141°C to 142°C | |
97.5% min. (GC) | |
1.4150 to 1.417 | |
0.893 | |
QKBBTQJLUGADEG-UHFFFAOYSA-N | |
3-(1-ethoxyethoxy)prop-1-yne | |
86774 | |
98% | |
Acetaldehyde ethyl propargyl acetal |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 242-490-
- Acetaldehyde ethyl propargyl acetal
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only