missing translation for 'onlineSavingsMsg'
Learn More

Acetaldoxime, 99%, mixture of syn and anti, Thermo Scientific Chemicals

£32.56 - £95.37

Chemical Identifiers

CAS 107-29-9
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.07
MDL Number MFCD00002124 MFCD00002124
InChI Key FZENGILVLUJGJX-NSCUHMNNSA-N
Synonym acetaldoxime, z-acetaldehyde oxime, 1z-acetaldehyde oxime, acetaldehyde, oxime, 1z, 1z-ethanal oxime, nz-n-ethylidenehydroxylamine, acetaldehyde, oxime, acetaldehyde, oxime, z, z-acetaldoxime, cis-acetaldehyde oxime
PubChem CID 5324280
ChEBI CHEBI:50719
IUPAC Name (NZ)-N-ethylidenehydroxylamine
SMILES C\C=N\O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10346210
View Documents
Thermo Scientific Acros
102230250
25 g Glass bottle
£32.56
25g
Please to purchase this item. Need a web account? Register with us today!
10162460
View Documents Promotion Details
Thermo Scientific Acros
102231000
100 g Glass bottle
£95.37
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

107-29-9
59.07
FZENGILVLUJGJX-NSCUHMNNSA-N
5324280
(NZ)-N-ethylidenehydroxylamine
C2H5NO
MFCD00002124 MFCD00002124
acetaldoxime, z-acetaldehyde oxime, 1z-acetaldehyde oxime, acetaldehyde, oxime, 1z, 1z-ethanal oxime, nz-n-ethylidenehydroxylamine, acetaldehyde, oxime, acetaldehyde, oxime, z, z-acetaldoxime, cis-acetaldehyde oxime
CHEBI:50719
C\C=N\O

Specifications

107-29-9
100.0
White
115.0°C
Authentic
C2H5NO
1.4250 to 1.4270
01, IV, 3121
15, 43
acetaldoxime, z-acetaldehyde oxime, 1z-acetaldehyde oxime, acetaldehyde, oxime, 1z, 1z-ethanal oxime, nz-n-ethylidenehydroxylamine, acetaldehyde, oxime, acetaldehyde, oxime, z, z-acetaldoxime, cis-acetaldehyde oxime
FZENGILVLUJGJX-NSCUHMNNSA-N
(NZ)-N-ethylidenehydroxylamine
5324280
59.06
Liquid or Low Melting Solid
98.5
44.0°C to 46.0°C
0.9600g/mL
38°C
98.5% min. (GC)
CH3CH=NOH
MFCD00002124 MFCD00002124
01,3
0.96
Solubility in water: soluble in water. Other solubilities: decomposes by aq. hcl,soluble in alcohol and ether
C\C=N\O
59.07
CHEBI:50719
99%
Acetaldoxime, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Flammable solid.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 203-479-6

missing translation for 'rtecsNumber' : AB2975000

missing translation for 'tsca' : TSCA

Product Identifier
  • Acetaldoxime, mixture of syn and anti

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Flammable solid Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only