Learn More
Acetaldoxime, syn + anti, 98%, Thermo Scientific Chemicals
Description
Acetaldoxime is used as an oxygen scavenger in boiler water. It is also used as an intermediate in chemical synthesis and pharmaceuticals. It is involved in the rearrangement reaction to prepare acetamide by using nickel(II) acetate as a catalyst. It acts as a precursor to prepare heterocyclic compound such as spiroisoxazoline. Further, it is used to prepare alkylated(Z)-oximes by deprotonation followed by reaction with benzyl bromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acetaldoxime is used as an oxygen scavenger in boiler water. It is also used as an intermediate in chemical synthesis and pharmaceuticals. It is involved in the rearrangement reaction to prepare acetamide by using nickel(II) acetate as a catalyst. It acts as a precursor to prepare heterocyclic compound such as spiroisoxazoline. Further, it is used to prepare alkylated(Z)-oximes by deprotonation followed by reaction with benzyl bromide.
Solubility
Soluble in water, ethanol and ether.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Density | 0.969 |
Boiling Point | 115°C to 117°C |
Flash Point | 36°C (96°F) |
Odor | Characteristic |
Refractive Index | 1.426 |
Quantity | 50 g |
UN Number | UN2332 |
Beilstein | 1209252 |
Merck Index | 14,42 |
Solubility Information | Soluble in water,ethanol and ether. |
Show More |
RUO – Research Use Only