missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetamidine hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.
Solubility
Soluble in water.
Notes
It is hygroscopic. Store away from oxidizing agents.
Acetamidine hydrochloride was used in the preparation of decarboxyectoine. It was also used in the synthesis of ethyl 4-(4-hydroxyphenyl)methylidene-2-methyl-5-oxo-1-imidazolacetate.
Solubility
Soluble in water.
Notes
It is hygroscopic. Store away from oxidizing agents.
Specifications
Melting Point | 168°C to 175°C |
Quantity | 25 g |
Beilstein | 3591762 |
Sensitivity | Hygroscopic |
Merck Index | 14,44 |
Solubility Information | Soluble in water. |
Formula Weight | 94.54 |
Percent Purity | 97% |
Chemical Name or Material | Acetamidine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'