missing translation for 'onlineSavingsMsg'
Learn More

Acetaminophen, Tocris Bioscience™

Catalog Number 18792584
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cyclooxygenase inhibitor; may be selective for COX-3

Cyclooxygenase inhibitor; may be selective for COX-3 (IC50 values are 460, > 1000 and > 1000 μM for canine COX-3, and murine COX-1 and COX-2 respectively). Widely used analgesic and antipyretic agent.
CAS 103-90-2
Molecular Formula C8H9NO2
MDL Number MFCD00002328
Quantity 100 mg
Synonym acetaminophen, 4-acetamidophenol, paracetamol, apap, n-4-hydroxyphenyl acetamide, tylenol, acetaminofen, panadol, datril, p-acetamidophenol
InChI Key RZVAJINKPMORJF-UHFFFAOYSA-N
SMILES CC(=O)NC1=CC=C(O)C=C1
IUPAC Name N-(4-hydroxyphenyl)acetamide
Molecular Weight (g/mol) 151.17
PubChem CID 1983
ChEBI CHEBI:46195
Formula Weight 151.16
Percent Purity >99%
Chemical Name or Material Acetaminophen
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'