missing translation for 'onlineSavingsMsg'
Learn More
Acetanilide, 98%, Thermo Scientific Chemicals Product Code.: 11374619

Acetanilide, 98%, Thermo Scientific Chemicals

Product Code. 11374619
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11374619

Brand: Thermo Scientific Alfa Aesar A14361.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acetanilide is used as an intermediate in the synthesis of rubber accelerator, dyes and camphor. It is also used in the synthesis of penicillin and other pharmaceutical products. It is involved in the preparation of 4-acetamidobenzenesulfonyl chloride, which is an intermediate during the synthesis of sulfa drugs. Further, it is employed as a experimental photographic developer. In addition to this, it is used to stabilize cellulose ester varnishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetanilide is used as an intermediate in the synthesis of rubber accelerator, dyes and camphor. It is also used in the synthesis of penicillin and other pharmaceutical products. It is involved in the preparation of 4-acetamidobenzenesulfonyl chloride, which is an intermediate during the synthesis of sulfa drugs. Further, it is employed as a experimental photographic developer. In addition to this, it is used to stabilize cellulose ester varnishes.

Solubility
Soluble in ethanol, diethyl ether, acetone, benzene. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-84-4
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00008674
InChI Key FZERHIULMFGESH-UHFFFAOYSA-N
Synonym acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene
PubChem CID 904
ChEBI CHEBI:28884
IUPAC Name N-phenylacetamide
SMILES CC(=O)NC1=CC=CC=C1

Specifications

Melting Point 113°C to 116°C
Density 1.121
Boiling Point 304°C to 305°C
Flash Point 161°C (321°F)
Assay Percent Range 98%
Quantity 100 g
Beilstein 606468
Merck Index 14,50
Solubility Information Soluble in ethanol,diethyl ether,acetone,benzene. Slightly soluble in water.
Formula Weight 135.17
Percent Purity 98%
Chemical Name or Material Acetanilide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248003 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog NumberA14361.22,A14361.36,A14361.0E
Lot Number10238195 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14361.22,A14361.36,A14361.0E
Lot Number10172904 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14361.22,A14361.36,A14361.0E
Lot Number10197739 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14361.22,A14361.36,A14361.0E
Lot Number10202623 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14361.22,A14361.36,A14361.0E

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetanilide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.