Learn More
Acetanilide, 99+%, Thermo Scientific Chemicals
£17.92 - £209.04
Chemical Identifiers
CAS | 103-84-4 |
---|---|
Molecular Formula | C8H9NO |
Molecular Weight (g/mol) | 135.17 |
MDL Number | MFCD00008674 |
InChI Key | FZERHIULMFGESH-UHFFFAOYSA-N |
Synonym | acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene |
PubChem CID | 904 |
ChEBI | CHEBI:28884 |
IUPAC Name | N-phenylacetamide |
SMILES | CC(=O)NC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10123440
|
Thermo Scientific Acros
150810050 |
5 g | Glass bottle |
£17.92
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10103490
|
Thermo Scientific Acros
150812500 |
250 g | Plastic bottle |
£22.05
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10091670
|
Thermo Scientific Acros
150815000 |
500 g | Plastic bottle |
£30.95
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10766381
|
Thermo Scientific Acros
150810010 |
1 kg | Plastic bottle |
£50.65
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10445755
|
Thermo Scientific Acros
150810051 |
5 kg | Plastic drum |
£209.04
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
103-84-4 | |
135.17 | |
FZERHIULMFGESH-UHFFFAOYSA-N | |
904 | |
N-phenylacetamide |
C8H9NO | |
MFCD00008674 | |
acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene | |
CHEBI:28884 | |
CC(=O)NC1=CC=CC=C1 |
Specifications
103-84-4 | |
100.0 | |
Blue-Gray to White | |
304.0°C | |
≥111°C to ≤115°C | |
99% min. (GC) | |
CH3CONHC6H5 | |
12, 237 | |
acetanilide, acetanil, acetamidobenzene, antifebrin, acetylaniline, acetanilid, n-acetylaniline, acetamide, n-phenyl, acetic acid anilide, acetylaminobenzene | |
FZERHIULMFGESH-UHFFFAOYSA-N | |
N-phenylacetamide | |
904 | |
135.17 | |
99+% | |
Acetanilide |
99.0 | |
111.0°C to 115.0°C | |
4.65 (Air = 1.0) | |
173°C | |
Conforms to Reference | |
C8H9NO | |
MFCD00008674 | |
15, 49 | |
Solubility in water: slightly soluble (5g/L at 25°C). Other solubilities: 1g/20mL boiling water,1g/3.4mL alcohol,1g/4mL ac,etone,1g/3.7mL chloroform,1g/5mL glycerol,1g/8mL,dioxane,1g/18mL diethyl ether,1g/47mL benzene,,sparingly soluble in pe | |
CC(=O)NC1=CC=CC=C1 | |
135.17 | |
CHEBI:28884 | |
13hPa at 114°C | |
Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-150-7
- Acetanilide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.