missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Acetate Buffer Solution Ph 4.6, Honeywell Fluka™

Product Code. 15643190 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500mL
1 L
6 x 500mL
Unit Size:
1 L
500mL
Pack of 6
This item is not returnable. View return policy

Product Code. 15643190

Brand: Honeywell Fluka™ 310486X500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sodium acetate / acetic acid, for complexometry

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 126-96-5
Product Type Acetate Buffer Solution Ph 4.6
Molecular Weight (g/mol) 143.094
Quantity 6 x 500mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.