missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACETATE BUFFER SOLUTION PH 4.65, SODIUM ACETATE / ACETIC ACID, Honeywell Fluka™
SODIUM ACETATE / ACETIC ACID
Brand: Honeywell-Fluka 31103-1L
39.03 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
127-09-3 | |
C2H3NaO2 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium acetate |
1 L | |
MFCD00012459 | |
[Na+].CC([O-])=O | |
82.03 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
29Hours
2Minutes
5Seconds
24 -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
21Minutes
54Seconds
35
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ACETATE BUFFER SOLUTION PH 4.65, SODIUM ACETATE / ACETIC ACID, Honeywell Fluka™ > 1L PLASTIC BOTTLE
Spot an opportunity for improvement?Share a Content Correction