missing translation for 'onlineSavingsMsg'
Learn More

Acetate solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™
BIOPHARMA

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

£49.50 - £104.00

Chemical Identifiers

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12942075
BIOPHARMA
View Documents
Fisher Chemical
A 002 250
250 mL Glass Bottle
£70.70
250mL
Please to purchase this item. Need a web account? Register with us today!
12952075
BIOPHARMA
View Documents
Fisher Chemical
A 002 500
500 mL Glass Bottle
£104.00
500mL
Please to purchase this item. Need a web account? Register with us today!
12932075
BIOPHARMA
View Documents
Fisher Chemical
A 002 100
500 mL
£49.50
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

127-09-3
82.03
VMHLLURERBWHNL-UHFFFAOYSA-M
[Na+].CC([O-])=O
C2H3NaO2
MFCD00012459
sodium acetate

Specifications

127-09-3
Liquid
1 g/L
[Na+].CC([O-])=O
Single-component Standard Solution
H2O
C2H3NaO2
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate
82.03
Acetate
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months. (12 Months after opening)

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetate solution for Ion Chromatography 1g/L H2O matrix, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.