missing translation for 'onlineSavingsMsg'
Learn More
Acetic Acid, 30%, European Pharmacopoiea Grade, Fisher Chemical™ Product Code.: 12997143
compliance-icons

Acetic Acid, 30%, European Pharmacopoiea Grade, Fisher Chemical™
BIOPHARMA

Product Code. 12997143
1000 L
Click to view available options
Quantity:
1000 L
This item is not returnable. View return policy

Product Code. 12997143

Brand: Fisher Chemical RP 001 1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Concentration 30%
Quantity 1000 L

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1092918 Certificate TypeCertificate of Analysis Date30/03/2025 Catalog NumberRP 001 1000
Lot Number2503001 Certificate TypeCertificate of Analysis Date25/03/2025 Catalog NumberRP 001 1000
Lot Number1093179 Certificate TypeCertificate of Analysis Date16/03/2025 Catalog NumberRP 001 1000
Lot Number1078841 Certificate TypeCertificate of Analysis Date05/03/2025 Catalog NumberRP 001 1000
Lot Number1002843 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 001 1000
compliance-icons
Product Identifier
  • Acetic acid,30%. European Pharmacopoeia Ref: 1000401 (RP 001)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetic Acid, 30%, European Pharmacopoiea Grade, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.