missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, 99.8%, for analysis, Thermo Scientific Chemicals

Product Code. 10542032
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
20 L
Packaging:
Glass bottle
Plastic Can
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 16.0°C to 16.5°C
Boiling Point 117.0°C to 118.0°C
CAS Min % 99.75
CAS Max % 100.0
Assay Percent Range 99.8%
Linear Formula CH3COOH
Beilstein 02, 96
Fieser 02,5; 05,3; 07,1; 08,1; 12,568; 13,45; 15,12
Merck Index 15, 54
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, glycerol, ether and, carbon tetrachloride, practically insoluble in carbon disulfide
Formula Weight 60.05
Percent Purity 99.8%
Grade Analytical
Density 1.0480g/mL
Quantity 20 L
Flash Point 40°C
Packaging Plastic Can
Physical Form Liquid
Refractive Index 1.3700 to 1.3730
Residue after Evaporation 0.001% max.
Specific Gravity 1.048
Chemical Name or Material Acetic acid, For Analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.