missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, ammonium salt, 98+%, for analysis, Thermo Scientific Chemicals

£43.92 - £239.19

Chemical Identifiers

CAS 631-61-8
Molecular Formula C2H7NO2
Molecular Weight (g/mol) 77.083
MDL Number MFCD00013066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
Synonym ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
PubChem CID 517165
ChEBI CHEBI:62947
IUPAC Name azanium;acetate
SMILES CC(=O)[O-].[NH4+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10543961
View Documents
Thermo Scientific
218365000
500 g Plastic bottle
£43.92
500g
Please to purchase this item. Need a web account? Register with us today!
10772641
View Documents
Thermo Scientific Acros
218360010
1 kg Plastic bottle
£69.55
1kg
Please to purchase this item. Need a web account? Register with us today!
10474795
View Documents
Thermo Scientific Acros
218360050
5 kg Plastic drum
£239.19
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for analysis
Specifications

Chemical Identifiers

631-61-8
77.083
USFZMSVCRYTOJT-UHFFFAOYSA-N
517165
azanium;acetate
C2H7NO2
MFCD00013066
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
CHEBI:62947
CC(=O)[O-].[NH4+]

Specifications

6.5 to 7.5 (5% soln. at 20°C)
114.0°C
100.0
98+%
CH3CO2NH4
02, 96
15, 492
Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C)
CC(=O)[O-].[NH4+]
77.083
CHEBI:62947
≥98%
136°C
Crystals or Crystalline Powder
631-61-8
98.0
White
C2H7NO2
MFCD00013066
01,38; 07,11
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4
USFZMSVCRYTOJT-UHFFFAOYSA-N
azanium;acetate
517165
77.08
Analytical
Authentic
Acetic acid, ammonium salt, Anhydrous
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Acetic acid, ammonium salt

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.