missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Acetic acid, ammonium salt, 98%, for biochemistry
£19.50 - £163.82
Chemical Identifiers
CAS | 631-61-8 |
---|---|
Molecular Formula | C2H7NO2 |
Molecular Weight (g/mol) | 77.083 |
InChI Key | USFZMSVCRYTOJT-UHFFFAOYSA-N |
Synonym | ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 |
PubChem CID | 517165 |
ChEBI | CHEBI:62947 |
IUPAC Name | azanium;acetate |
SMILES | CC(=O)[O-].[NH4+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12397183
|
Thermo Scientific Chemicals
447121000 |
100 g | Plastic bottle |
£19.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12307193
|
Thermo Scientific Chemicals
447125000 |
500 g | Plastic bottle |
£61.61
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12387183
|
Thermo Scientific Chemicals
447120025 |
2.5 kg | Plastic bottle |
£163.82
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistryChemical Identifiers
631-61-8 | |
77.083 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CHEBI:62947 | |
CC(=O)[O-].[NH4+] |
C2H7NO2 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
517165 | |
azanium;acetate |
Specifications
(c=1M, water), (c=1M, water) at 0nm, 0.01 max. at 280nm, 0.01 max. at 350nm, 0.02 max. at 254nm | |
114.0°C | |
C2H7NO2 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
azanium;acetate | |
517165 | |
77.08 | |
Biochemical | |
Acetic acid, ammonium salt |
631-61-8 | |
98% | |
CH3CO2NH4 | |
Solubility in water: 1480g/L (20°C). Other solubilities: soluble in alcohol, slightly soluble in acetone, in methanol: 7.89 g/100mL (15°C) | |
CC(=O)[O-].[NH4+] | |
77.083 | |
CHEBI:62947 | |
98% | |
136°C |
Safety and Handling
Product Identifier
- Acetic acid, ammonium salt
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title