missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, ammonium salt-d7, 99 atom % D, Thermo Scientific Chemicals
£37.71 - £117.00
Chemical Identifiers
CAS | 194787-05-8 |
---|---|
Molecular Formula | C2H7NO2 |
Molecular Weight (g/mol) | 84.126 |
InChI Key | USFZMSVCRYTOJT-KYWLLVRESA-N |
Synonym | ammonium acetate-d7, 98 atom % d, acetic-d3 acid-d,ammonium-d4 salt 9ci, 2 h? amine; 2 h? ethan 2 h oic acid, ∼2∼h_4_ acetic acid-∼2∼h_3_ ammonia 1/1 |
PubChem CID | 51341917 |
SMILES | CC(=O)O.N |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
194787-05-8 | |
84.126 | |
ammonium acetate-d7, 98 atom % d, acetic-d3 acid-d,ammonium-d4 salt 9ci, 2 h? amine; 2 h? ethan 2 h oic acid, ∼2∼h_4_ acetic acid-∼2∼h_3_ ammonia 1/1 | |
CC(=O)O.N |
C2H7NO2 | |
USFZMSVCRYTOJT-KYWLLVRESA-N | |
51341917 |
Specifications
Glass Bottle | |
C2H7NO2 | |
ammonium acetate-d7, 98 atom % d, acetic-d3 acid-d,ammonium-d4 salt 9ci, 2 h? amine; 2 h? ethan 2 h oic acid, ∼2∼h_4_ acetic acid-∼2∼h_3_ ammonia 1/1 | |
CC(=O)O.N | |
51341917 | |
Acetic acid, ammonium salt-d7 |
194787-05-8 | |
CD3CO2ND4 | |
USFZMSVCRYTOJT-KYWLLVRESA-N | |
84.126 | |
84.1 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetic acid, ammonium salt-d7
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only